Anti-NDUFC2 Rabbit Polyclonal Antibody

Supplier: Abnova
H00004718-D01P
10594-598EA 376 USD
10594-598
Anti-NDUFC2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFC2 (NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR