Anti-NDUFB10 Rabbit Polyclonal Antibody

Supplier: Abnova
H00004716-D01
10594-576EA 376 USD
10594-576
Anti-NDUFB10 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFB10 (NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10, 22kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR