Anti-NDUFB1 Mouse Polyclonal Antibody

H00004707-A01
10565-440EA 243 USD
10565-440
Anti-NDUFB1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFB1 (NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 1, 7kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR