Anti-NDUFAB1 Mouse Monoclonal Antibody [clone: 2D10]

Supplier: Abnova
H00004706-M06
10594-570EA 335 USD
10594-570
Anti-NDUFAB1 Mouse Monoclonal Antibody [clone: 2D10]
Antibodies
Type: Primary
Antigen: NDUFAB1 (NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1, 8kDa)
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR