Anti-NEB Mouse Monoclonal Antibody [clone: 1F4]

Supplier: Abnova
H00004703-M01
10594-614EA 335 USD
10594-614
Anti-NEB Mouse Monoclonal Antibody [clone: 1F4]
Antibodies
Type: Primary
Antigen: NEB (nebulin)
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR