Anti-NDUFA5 Mouse Polyclonal Antibody

Supplier: Abnova
H00004698-B01
10565-386EA 335 USD
10565-386
Anti-NDUFA5 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFA5 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR