Anti-NDUFA1 Mouse Monoclonal Antibody [clone: 2A4]

Supplier: Abnova
H00004694-M10
10594-552EA 335 USD
10594-552
Anti-NDUFA1 Mouse Monoclonal Antibody [clone: 2A4]
Antibodies
Type: Primary
Antigen: NDUFA1 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1, 7.5kDa)
Clonality: Monoclonal
Clone: 2A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR