Anti-NCK1 Mouse Monoclonal Antibody [clone: 1A1]

Supplier: Abnova
H00004690-M01
10594-484EA 335 USD
10594-484
Anti-NCK1 Mouse Monoclonal Antibody [clone: 1A1]
Antibodies
Type: Primary
Antigen: NCK1
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR