Anti-NCK1 Mouse Polyclonal Antibody

H00004690-B01
10565-316EA 335 USD
10565-316
Anti-NCK1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR