Anti-NAB2 Mouse Monoclonal Antibody [clone: 4H6]

Supplier: Abnova
H00004665-M01
10594-382EA 335 USD
10594-382
Anti-NAB2 Mouse Monoclonal Antibody [clone: 4H6]
Antibodies
Type: Primary
Antigen: NAB2
Clonality: Monoclonal
Clone: 4H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR