Anti-MYL5 Mouse Monoclonal Antibody [clone: 1C5]

Supplier: Abnova
H00004636-M01
10594-176EA 335 USD
10594-176
Anti-MYL5 Mouse Monoclonal Antibody [clone: 1C5]
Antibodies
Type: Primary
Antigen: MYL5
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR