Anti-MYH9 Mouse Monoclonal Antibody [clone: 4H3]

Supplier: Abnova
H00004627-M06
10594-162EA 335 USD
10594-162
Anti-MYH9 Mouse Monoclonal Antibody [clone: 4H3]
Antibodies
Type: Primary
Antigen: MYH9
Clonality: Monoclonal
Clone: 4H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR