Anti-MYH9 Mouse Monoclonal Antibody [clone: 4D1]

Supplier: Abnova
H00004627-M04
10594-158EA 335 USD
10594-158
Anti-MYH9 Mouse Monoclonal Antibody [clone: 4D1]
Antibodies
Type: Primary
Antigen: MYH9
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR