Anti-MYBPH Mouse Monoclonal Antibody [clone: 1G18]

Supplier: Abnova
H00004608-M02A
10625-370EA 335 USD
10625-370
Anti-MYBPH Mouse Monoclonal Antibody [clone: 1G18]
Antibodies
Type: Primary
Antigen: MYBPH
Clonality: Monoclonal
Clone: 1G18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR