Anti-MUSK Mouse Monoclonal Antibody [clone: 4C12]

Supplier: Abnova
H00004593-M03
10594-074EA 335 USD
10594-074
Anti-MUSK Mouse Monoclonal Antibody [clone: 4C12]
Antibodies
Type: Primary
Antigen: MUSK
Clonality: Monoclonal
Clone: 4C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR