Anti-MUSK Mouse Polyclonal Antibody

H00004593-A01
10564-950EA 243 USD
10564-950
Anti-MUSK Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MUSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR