Anti-TRIM37 Mouse Polyclonal Antibody

H00004591-A01
10572-946EA 243 USD
10572-946
Anti-TRIM37 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRIM37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR