Anti-MTF1 Mouse Monoclonal Antibody [clone: 2C12]

Supplier: Abnova
H00004520-M04
10593-982EA 335 USD
10593-982
Anti-MTF1 Mouse Monoclonal Antibody [clone: 2C12]
Antibodies
Type: Primary
Antigen: MTF1
Clonality: Monoclonal
Clone: 2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR