Anti-MSRA Mouse Monoclonal Antibody [clone: 3C11]

Supplier: Abnova
H00004482-M01
10593-912EA 325 USD
10593-912
Anti-MSRA Mouse Monoclonal Antibody [clone: 3C11]
Type: Primary
Antigen: MSRA
Clonality: Monoclonal
Clone: 3C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR