Anti-MSR1 Mouse Monoclonal Antibody [clone: 2G8]

Supplier: Abnova
H00004481-M01
10593-908EA 335 USD
10593-908
Anti-MSR1 Mouse Monoclonal Antibody [clone: 2G8]
Antibodies
Type: Primary
Antigen: MSR1
Clonality: Monoclonal
Clone: 2G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR