Anti-MSI1 Mouse Monoclonal Antibody [clone: 4A11]

Supplier: Abnova
H00004440-M06
10593-884EA 335 USD
10593-884
Anti-MSI1 Mouse Monoclonal Antibody [clone: 4A11]
Antibodies
Type: Primary
Antigen: MSI1
Clonality: Monoclonal
Clone: 4A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR