Anti-MSH2 Mouse Monoclonal Antibody [clone: 4F11]

Supplier: Abnova
H00004436-M04
10593-872EA 335 USD
10593-872
Anti-MSH2 Mouse Monoclonal Antibody [clone: 4F11]
Antibodies
Type: Primary
Antigen: MSH2
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR