Anti-MSH2 Mouse Monoclonal Antibody [clone: 4B2]

Supplier: Abnova
H00004436-M02
10593-866EA 335 USD
10593-866
Anti-MSH2 Mouse Monoclonal Antibody [clone: 4B2]
Antibodies
Type: Primary
Antigen: MSH2
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR