Anti-CITED1 Mouse Monoclonal Antibody [clone: 2H6]

Supplier: Abnova
H00004435-M06
10582-280EA 335 USD
10582-280
Anti-CITED1 Mouse Monoclonal Antibody [clone: 2H6]
Antibodies
Type: Primary
Antigen: CITED1
Clonality: Monoclonal
Clone: 2H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR