Anti-CITED1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00004435-D01
10582-282EA 376 USD
10582-282
Anti-CITED1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CITED1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR