Anti-MRE11A Mouse Monoclonal Antibody [clone: 3H4-F4]

Supplier: Abnova
H00004361-M02
10593-792EA 335 USD
10593-792
Anti-MRE11A Mouse Monoclonal Antibody [clone: 3H4-F4]
Antibodies
Type: Primary
Antigen: MRE11A
Clonality: Monoclonal
Clone: 3H4-F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR