Anti-MRC1 Mouse Monoclonal Antibody [clone: 5C11]

Supplier: Abnova
H00004360-M02
10593-690EA 335 USD
10593-690
Anti-MRC1 Mouse Monoclonal Antibody [clone: 5C11]
Antibodies
Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: 5C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR