Anti-MAP3K9 Mouse Polyclonal Antibody

Supplier: Abnova
H00004293-A01
10563-722EA 243 USD
10563-722
Anti-MAP3K9 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP3K9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR