Anti-CD99 Mouse Monoclonal Antibody [clone: 3A10]

Supplier: Abnova
H00004267-M01
10581-560EA 335 USD
10581-560
Anti-CD99 Mouse Monoclonal Antibody [clone: 3A10]
Antibodies
Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: 3A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR