Anti-MGST2 Mouse Monoclonal Antibody [clone: 1D11]

Supplier: Abnova
H00004258-M01A
10624-660EA 335 USD
10624-660
Anti-MGST2 Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
Type: Primary
Antigen: MGST2
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR