Anti-KITLG Mouse Monoclonal Antibody [clone: 2A10]

Supplier: Abnova
H00004254-M04
10591-150EA 335 USD
10591-150
Anti-KITLG Mouse Monoclonal Antibody [clone: 2A10]
Antibodies
Type: Primary
Antigen: KITLG
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR