Anti-MFAP3 Mouse Monoclonal Antibody [clone: 1C7]

Supplier: Abnova
H00004238-M01
10593-262EA 335 USD
10593-262
Anti-MFAP3 Mouse Monoclonal Antibody [clone: 1C7]
Antibodies
Type: Primary
Antigen: MFAP3
Clonality: Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR