Anti-MEOX2 Mouse Monoclonal Antibody [clone: 4B2]

Supplier: Abnova
H00004223-M11
10593-234EA 335 USD
10593-234
Anti-MEOX2 Mouse Monoclonal Antibody [clone: 4B2]
Antibodies
Type: Primary
Antigen: MEOX2
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR