Anti-MAP3K4 Mouse Monoclonal Antibody [clone: 4F10]

Supplier: Abnova
H00004216-M08
10591-878EA 335 USD
10591-878
Anti-MAP3K4 Mouse Monoclonal Antibody [clone: 4F10]
Antibodies
Type: Primary
Antigen: MAP3K4
Clonality: Monoclonal
Clone: 4F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR