Anti-MAP3K4 Mouse Polyclonal Antibody

Supplier: Abnova
H00004216-A01
10563-700EA 243 USD
10563-700
Anti-MAP3K4 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP3K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR