Anti-MEFV Mouse Monoclonal Antibody [clone: 2C1]

Supplier: Abnova
H00004210-M02
10593-200EA 335 USD
10593-200
Anti-MEFV Mouse Monoclonal Antibody [clone: 2C1]
Antibodies
Type: Primary
Antigen: MEFV
Clonality: Monoclonal
Clone: 2C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR