Anti-MEF2D Mouse Monoclonal Antibody [clone: 3F8]

Supplier: Abnova
H00004209-M04
10593-194EA 335 USD
10593-194
Anti-MEF2D Mouse Monoclonal Antibody [clone: 3F8]
Antibodies
Type: Primary
Antigen: MEF2D
Clonality: Monoclonal
Clone: 3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR