Anti-INPPL1 Mouse Monoclonal Antibody [clone: 10G6]

Supplier: Abnova
H00003636-M02
10590-558EA 335 USD
10590-558
Anti-INPPL1 Mouse Monoclonal Antibody [clone: 10G6]
Antibodies
Type: Primary
Antigen: INPPL1
Clonality: Monoclonal
Clone: 10G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR