Anti-IDO1 Mouse Monoclonal Antibody [clone: 3D12-C6]

Supplier: Abnova
H00003620-M01A
10624-520EA 325 USD
10624-520
Anti-IDO1 Mouse Monoclonal Antibody [clone: 3D12-C6]
Antibodies
Type: Primary
Antigen: IDO1
Clonality: Monoclonal
Clone: 3D12-C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR