Anti-IL15 Mouse Monoclonal Antibody [clone: 3A3]

Supplier: Abnova
H00003600-M06
10590-142EA 335 USD
10590-142
Anti-IL15 Mouse Monoclonal Antibody [clone: 3A3]
Antibodies
Type: Primary
Antigen: IL15
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR