Anti-CXCL8 Mouse Monoclonal Antibody [clone: 4G10]

Supplier: Abnova
H00003576-M07
10590-376EA 335 USD
10590-376
Anti-CXCL8 Mouse Monoclonal Antibody [clone: 4G10]
Antibodies
Type: Primary
Antigen: CXCL8
Clonality: Monoclonal
Clone: 4G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR