Anti-IL4R Mouse Monoclonal Antibody [clone: 3E5]

Supplier: Abnova
H00003566-M04A
10624-528EA 335 USD
10624-528
Anti-IL4R Mouse Monoclonal Antibody [clone: 3E5]
Antibodies
Type: Primary
Antigen: IL4R
Clonality: Monoclonal
Clone: 300000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR