Anti-CD40L Mouse Monoclonal Antibody [clone: 2G3]

Supplier: Abnova
H00000959-M05
10571-320EA 335 USD
10571-320
Anti-CD40L Mouse Monoclonal Antibody [clone: 2G3]
Antibodies
Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 2G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR