Anti-CD40L Mouse Polyclonal Antibody

Supplier: Abnova
H00000959-A01
10572-666EA 243 USD
10572-666
Anti-CD40L Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD40L
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR