Anti-ENTPD5 Mouse Monoclonal Antibody [clone: 4A5]

Supplier: Abnova
H00000957-M07
10585-046EA 335 USD
10585-046
Anti-ENTPD5 Mouse Monoclonal Antibody [clone: 4A5]
Antibodies
Type: Primary
Antigen: ENTPD5
Clonality: Monoclonal
Clone: 4A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR