Anti-CD34 Mouse Monoclonal Antibody [clone: 5F3]

Supplier: Abnova
H00000947-M01
10579-080EA 335 USD
10579-080
Anti-CD34 Mouse Monoclonal Antibody [clone: 5F3]
Antibodies
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 5F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR