Anti-CD33 Mouse Polyclonal Antibody

H00000945-B01P
10552-810EA 316.92 USD
10552-810
Anti-CD33 Mouse Polyclonal Antibody
Type: Primary
Antigen: CD33
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR