Anti-CD27 Rabbit Polyclonal Antibody

Supplier: Abnova
H00000939-D01P
10604-260EA 376 USD
10604-260
Anti-CD27 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR