Anti-CD19 Mouse Monoclonal Antibody [clone: 1G3]

Supplier: Abnova
H00000930-M01A
10623-996EA 335 USD
10623-996
Anti-CD19 Mouse Monoclonal Antibody [clone: 1G3]
Antibodies
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR