Anti-CD19 Rabbit Polyclonal Antibody

Supplier: Abnova
H00000930-D01P
10579-030EA 376 USD
10579-030
Anti-CD19 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR